• USA Home
  • N213 - [Phe1-ψ(CH2-NH)-Gly2]-Nociceptin Fragment 1-13 amide

EMAIL THIS PAGE TO A FRIEND
N213 Sigma

[Phe1-ψ(CH2-NH)-Gly2]-Nociceptin Fragment 1-13 amide

lyophilized powder

  • Empirical Formula (Hill Notation) C61H101N22O14

  • Molecular Weight 1366.59

  •  MDL number MFCD01631267

  •  PubChem Substance ID 329818596

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Opioid Peptides, Other Opioid Peptides,
InChI Key   ZHKMSRDIVOXQKP-YILJZHMHSA-N
form   lyophilized powder
storage temp.   −20°C
Gene Information   human ... PNOC(5368)

Description

Amino Acid Sequence

Phe-(CH2-NH)-Gly-Gly-Phe-Thr-Gly-Ala-Arg-Lys-Ser-Ala-Arg-Lys-NH2

Biochem/physiol Actions

Selective ORL1 opioid receptor antagonist at peripheral receptors. It functions as a partial agonist at central ORL1 receptors.

Price and Availability


Amplified Detection

Free Webinar

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?