EMAIL THIS PAGE TO A FRIEND
N215 Sigma

Nocistatin

>98%, lyophilized powder

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Opioid Peptides, Other Opioid Peptides,
assay   >98%
form   lyophilized powder
storage temp.   −20°C

Description

Amino Acid Sequence

Thr-Glu-Pro-Gly-Leu-Glu-Glu-Val-Gly-Glu-Ile-Glu-Gln-Lys-Gln-Leu-Gln

Biochem/physiol Actions

Endogenous 17 amino acid peptide from the orphanin FQ precursor, prepronociceptin/orphanin FQ, that does not bind to ORL1 receptors but is a functional antagonist of orphaninFQ/nociceptin.

Price and Availability


Making Headway

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?