• USA Home
  • N2268 - Nω-Nitro-L-arginine p-nitroanilide hydrobromide

EMAIL THIS PAGE TO A FRIEND
N2268 Sigma

Nω-Nitro-L-arginine p-nitroanilide hydrobromide

  • CAS Number 85697-89-8

  • Empirical Formula (Hill Notation) C12H17N7O5 · HBr

  • Molecular Weight 420.22

  •  MDL number MFCD01318756

  •  PubChem Substance ID 24897564

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   VLIUCLVWDVXVTH-PPHPATTJSA-N
impurities   ≤20% dioxane as solvent of crystallization
storage temp.   −20°C

Description

Packaging

100 mg in poly bottle

Biochem/physiol Actions

Nω-Nitro-L-arginine p-nitroanilide hydrobromide inhibits neuronal NO synthase.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?