• USA Home
  • N2394 - Nylon membranes, charge modified
EMAIL THIS PAGE TO A FRIEND
N2394 Sigma
Nylon membranes, charge modified
pore size 0.45 μm, size 15 cm × 15 cm
Purchase
Description
General description
Non-curling, positively charged Nylon 66 membrane tested for suitability in electroblotting of proteins. Also suitable for gold staining and immunoblotting. Anionic dyes (e.g., Amido Black, Coomassie Blue, Ponceau S) give high backgrounds with these membranes.
Price and Availability
Antibody Explorer
Amicon Pro Purification System
SNAP and Go!
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?