• USA Home
  • N2397 - Nitrate Reductase from corn

EMAIL THIS PAGE TO A FRIEND
N2397 Sigma

Nitrate Reductase from corn

buffered aqueous glycerol solution, ≥4 units/mL

Synonym: NADH:nitrate oxidoreductase

Purchase

Description

Application

Catalyzes the NADH-dependent reduction of nitrate to nitrite.

General description

Formerly E.C. 1.6.6.1

Unit Definition

One unit will reduce 1.0 μmole of nitrate to nitrite per minute in a NADH system at pH 7.3 at 30 °C.

Physical form

Solution in 0.01 M EDTA, 50 mM MOPS, pH 7.0, containing 50% glycerol and 1 mg per mL BSA

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?