• USA Home
  • N2505 - 4-Nitrophenyl β-L-fuco­pyran­oside

EMAIL THIS PAGE TO A FRIEND
N2505 Sigma

4-Nitrophenyl β-L-fuco­pyran­oside

Purchase

Properties

InChI Key   YILIDCGSXCGACV-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?