EMAIL THIS PAGE TO A FRIEND
N2884 Sigma

Nitromethane

ACS reagent, ≥95%

  • CAS Number 75-52-5

  • Empirical Formula (Hill Notation) CH3NO2

  • Molecular Weight 61.04

  •  Beilstein Registry Number 1698205

  •  EC Number 200-876-6

  •  MDL number MFCD00007400

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   ACS reagent
vapor density   2.1 (vs air)
InChI Key   LYGJENNIWJXYER-UHFFFAOYSA-N
assay   ≥95%
autoignition temp.   784 °F
expl. lim.   7.3 %, 33 °F
refractive index   n20/D 1.382(lit.)
bp   101.2 °C(lit.)
mp   −29 °C(lit.)
density   1.127 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1261 - class 3 - PG 2 - Nitromethane
WGK Germany 
2
RTECS 
PA9800000
Flash Point(F) 
96.8 °F
Flash Point(C) 
36 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?