• USA Home
  • N2905 - Naphthol AS-BI phosphate disodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
N2905 Sigma

Naphthol AS-BI phosphate disodium salt hydrate

BioXtra

Synonym: 2-Naphthalenecarboxamide, 7-bromo-N-(2-methoxyphenyl)-3-(phosphonooxy)-, disodium salt hydrate

  • CAS Number 207569-04-8

  • Empirical Formula (Hill Notation) C18H13BrNNa2O6P · xH2O

  • Molecular Weight 496.16 (anhydrous basis)

  •  PubChem Substance ID 24897605

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Histochemical (insoluble) More...
InChI Key   WNPKMQCBZKBNOS-UHFFFAOYSA-L
product line   BioXtra
assay   ≥99.5%
impurities   ≤0.1% Insoluble matter
solubility   H2O: soluble0.1 M at 20 °C, clear, colorless to faintly yellow
anion traces   sulfate (SO42-): ≤0.05%
cation traces   Al: ≤0.0005%
  Ca: ≤0.001%
  Cu: ≤0.0005%
  Fe: ≤0.001%
  K: ≤0.005%
  Mg: ≤0.002%
  Pb: ≤0.001%
  Zn: ≤0.0005%
storage temp.   −20°C

Description

Substrates

Substrate for the histochemical demonstration of acid and alkaline phosphatase.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?