• USA Home
  • N3015 - Hamilton® syringe needles
EMAIL THIS PAGE TO A FRIEND
N3015 Sigma
Hamilton® syringe needles
for series TLL (PTFE Luer-Lock) syringes, size × bevel 22 S gauge × 22 °, L 2 in.
Purchase
Properties
Related Categories Hamilton needles, Hamilton syringes, Labware, Syringes and Needles
L   2 in.
size × bevel   22 S gauge × 22 °
Description
General description
Hamilton syringes in Series N are provided with cemented needles that are not replaceable by the user. Series RN and KH syringes are provided with needles that are removable and user-replaceable. Needles for Series LT and TLL syringes must be purchased separately.
Replacement needles for RN syringes, and accessory needles for LT and TLL syringes are listed below. KH needles are listed after the Series 7000 syringes with which they are used.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?