• USA Home
  • N3513 - 4-Nitrophenyl α-L-rhamnopyranoside

EMAIL THIS PAGE TO A FRIEND
N3513 Sigma

4-Nitrophenyl α-L-rhamnopyranoside

~99% (TLC)

Synonym: p-Nitrophenyl 6-deoxy-α-L-mannopyranoside

  • CAS Number 18918-31-5

  • Empirical Formula (Hill Notation) C12H15NO7

  • Molecular Weight 285.25

  •  MDL number MFCD00069788

Purchase

Properties

InChI Key   YILIDCGSXCGACV-UHFFFAOYSA-N
assay   ~99% (TLC)
storage temp.   −20°C

Description

Substrates

Chromogenic substrate for naringinase.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

N7763 4-Nitrophenyl α-L-rhamnopyranoside, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?