• USA Home
  • N3521 - Filter membranes, nitrocellulose

EMAIL THIS PAGE TO A FRIEND
N3521 Sigma

Filter membranes, nitrocellulose

MF membrane, pore size 5 μm, diam. 13 mm

Purchase

Properties

Related Categories Filter membranes, nitrocellulose, Filtration Supplies, Labware, Manufacturer Browser, Membrane Filters,
material   MF membrane
mfr. no.   Millipore SMWP01300
  Millipore, SMWP01300
diam.   13 mm
pore size   5 μm

Description

General description

Biologically inert nitrocellulose membranes containing a small amount of cellulose acetate for improved handling. Autoclavable to 121°C; use below 75°C. Chemically compatible with dilute acids and bases, hydrocarbons, and non-polar liquids. MF-Millipore membranes have water extractables <5%, including a Triton wetting agent TF-Millipore membranes are Triton-free and have even lower water extractables than MF membranes. See Immobilon-NC in the Electrophoresis Equipment section for nitrocellulose blotting membranes.

Price and Availability


Sterifil Aseptic System and Holder
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?