• USA Home
  • N4034 - NCS-356 sodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
N4034 Sigma

NCS-356 sodium salt hydrate

solid, ≥98% (HPLC)

Synonym: γ-p-Chlorophenyl-trans-4-hydroxycrotonate sodium salt hydrate, 4-(4-Chlorophenyl)-4-hydroxy-2-butenoic acid sodium salt hydrate

  • CAS Number 430440-66-7

  • Empirical Formula (Hill Notation) C10H8ClO3Na · xH2O

  • Molecular Weight 234.61 (anhydrous basis)

  •  PubChem Substance ID 24278586

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, GABAergics,
InChI Key   NZFKWKOBJBRWMV-TXOOBNKBSA-M
assay   ≥98% (HPLC)
form   solid
color   white to off-white
solubility   H2O: ≥10 mg/mL

Description

Biochem/physiol Actions

γ-Hydroxybutyrate (GHB) receptor agonist.

Price and Availability


Neuroscience Grant Programm
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?