• USA Home
  • N4281 - BioBond Nylon transfer membrane
EMAIL THIS PAGE TO A FRIEND
N4281 Sigma
BioBond Nylon transfer membrane
disc size 132 mm
Purchase
Description
General description
• Controlled 0.45 μm pore size and high void volume for minimal resistance to flow and high diffusion rates.
• Allows for high sensitivity detection in both DNA and RNA blotting.
• Nucleic acids can be bound to the membrane by either UV cross-linking or baking.
• Inherent hydrophilicity ensures instantaneous water wetting; no pre-wetting needed.
• Specially gridded interleaf for accurate cutting of rolls and sheets.
Other Notes
This is a neutral, supported, untreated nylon membrane especially suited for non-radioactive Southern and Northern blotting, dot blots, and colony lifts.
Legal Information
BioBond is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?