• USA Home
  • N4520 - 2-Nitro-1H-imidazole-2-14C

EMAIL THIS PAGE TO A FRIEND
N4520 Sigma

2-Nitro-1H-imidazole-2-14C

~95% (Radiochemical Purity), methanol solution

Synonym: Azomycin

Purchase

Properties

assay   ~95% (Radiochemical Purity)
form   methanol solution
extent of labeling   ≥5 mCi per mmol
storage temp.   2-8°C

Description

Packaging

sealed ampule

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?