• USA Home
  • N4629 - 2-Naphthoxyacetic acid N-hydroxysuccinimide ester

EMAIL THIS PAGE TO A FRIEND
N4629 Sigma

2-Naphthoxyacetic acid N-hydroxysuccinimide ester

practical grade

Synonym: N-Succinimidyl 2-naphthoxyacetate

Purchase

Properties

grade   practical grade
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?