• USA Home
  • N5132 - N-Nitrosodibutylamine

EMAIL THIS PAGE TO A FRIEND
N5132 Sigma

N-Nitrosodibutylamine

Synonym: Dibutyl nitrosamine

Purchase

Properties

InChI Key   YGJHZCLPZAZIHH-UHFFFAOYSA-N
density   0.9 g/mL(lit.)

Description

Packaging

Bulk package

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
EJ4025000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

442685 N-Nitrosodi-n-butylamine, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?