• USA Home
  • N5135 - Normorphine hydrochloride monohydrate

EMAIL THIS PAGE TO A FRIEND
N5135 Sigma

Normorphine hydrochloride monohydrate

  • CAS Number 3372-02-9

  • Empirical Formula (Hill Notation) C16H17NO3 · HCl · H2O

  • Molecular Weight 325.79

  •  EC Number 222-152-9

  • Popular Documents:  Datasheet (PDF)  

Purchase

Properties

drug control   USDEA Schedule I; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

Description

Biochem/physiol Actions

Narcotic analgesic

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?