• USA Home
  • N5147 - Cytidine 3′:5′-cyclic monophosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
N5147 Sigma

Cytidine 3′:5′-cyclic monophosphate sodium salt

suitable for HPLC marker

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Biomarkers and Metabolomics Standards, Nucleotide & Nucleoside Standards
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for HPLC marker
format   neat
storage temp.   −20°C

Description

Components

Total nucleoside/nucleotide content is approximately 6 μmole, or 2 mg, per vial.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?