• USA Home
  • N5409 - Northern Bound Simian Monkey mRNA Blot
EMAIL THIS PAGE TO A FRIEND
N5409 Sigma
Northern Bound Simian Monkey mRNA Blot
Purchase
Description
General description
The Simian monkey blot allows for tissue specific gene expression studies in normal monkey tissues including, heart, brain, kidney, lung, spleen, skeletal muscle and small intestine. 3 μg of each source tissue mRNA is included on this 8-lane blot which also includes human lung tissue from normal adult as an interblot control for comparison between different blots.
Legal Information
Manufactured by BioChain, Inc.
Northern Bound is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?