• USA Home
  • N5643 - Nucleoside Phosphorylase from calf spleen

EMAIL THIS PAGE TO A FRIEND
N5643 Sigma

Nucleoside Phosphorylase from calf spleen

Synonym: Purine nucleoside phosphorylase, Purine nucleoside: orthophosphate ribosyltransferase

Purchase

Properties

Related Categories 2.4.x.x Glycosyltransferases, 2.x.x.x Transferases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
activity   1.5-3.5 units/mg solid
packaging   vial of 40-60 units
anion traces   phosphate (PO43-): <0.1%
storage temp.   2-8°C

Description

Unit Definition

One unit will cause the phosphorolysis of 1.0 μmole of inosine to hypoxanthine and ribose 1-phosphate per min at pH 7.4 at 25°C.

Physical form

Phosphate-free, lyophilized powder

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?