• USA Home
  • N5656 - Biobond-Plus Nylon transfer membrane
EMAIL THIS PAGE TO A FRIEND
N5656 Sigma
Biobond-Plus Nylon transfer membrane
sheet size 30 cm × 60 cm
Purchase
Description
General description
• Controlled 0.45 μm pore size and high void volume for minimal resistance to flow and high diffusion rates
• Superior signal-to-noise ratio for high sensitivity and extremely low background
• Strong DNA binding can be achieved without UV light or baking
• Specially gridded interleaf for accurate cutting of rolls and sheets
Other Notes
This is a positively charged, supported, derivatized nylon membrane especially suited for radioactive Southern and Northern blotting, dot blots, and colony lifts.
Legal Information
Biobond-Plus is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?