• USA Home
  • N5659 - Northern Bound Human Alzheimer’s Total RNA Blot II

EMAIL THIS PAGE TO A FRIEND
N5659 Sigma

Northern Bound Human Alzheimer’s Total RNA Blot II

from human

Purchase

Properties

Related Categories Gene Expression & Analysis, Molecular Biology, Multiple Tissue Northern Blots More...
biological source   from human

Description

General description

Blot includes 7 lanes of total RNA isolated from Alzheimer′s-diseased brain tissue including: frontal lobe, temporal lobe, parietal lobe, occipital lobe, pons, thalamus and corpus callosum.

Legal Information

Manufactured by BioChain, Inc.

Northern Bound is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?