• USA Home
  • N5783 - N-(NBD-Aminocaproyl)phytosphingosine

EMAIL THIS PAGE TO A FRIEND
N5783 Sigma

N-(NBD-Aminocaproyl)phytosphingosine

>98%

Synonym: C6-NBD Phytosphingosine, N-[6-[(7-Nitro-2-1,3-benzoxadiazol-4-yl)amino]caproyl]phytosphingosine

  • Empirical Formula (Hill Notation) C30H51N5O7

  • Molecular Weight 593.76

  •  MDL number MFCD02259242

  •  PubChem Substance ID 329818657

Purchase

Description

General description

Fluorescently labeled fatty acid sphingolipid membrane probe. NBD group excitation 460 nm, emission 534 nm.

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?