EMAIL THIS PAGE TO A FRIEND
N57 Sigma

Triton N-57

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Detergents N to Z, Non-Ionic Detergents More...
description   non-ionic

Description

General description

Triton N-57 is a surfactant with the ability to combat algal parasites Phlyctidium scenedesmi (Fott) and Phlyctidium sp.

Packaging

500 mL in poly bottle

Application

Triton N-57 has been used in a study to assess calcium activated neutral proteinase activity in brain and other tissue of rat. It has also been used in a study to synthesize fine silica powders from sodium silicate at room temperature.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?