• USA Home
  • N5887 - 3-(1-Naphthyl)-D-alanine

EMAIL THIS PAGE TO A FRIEND
N5887 Sigma

3-(1-Naphthyl)-D-alanine

  • CAS Number 78306-92-0

  • Empirical Formula (Hill Notation) C13H13NO2

  • Molecular Weight 215.25

  •  Beilstein Registry Number 4352834

  •  MDL number MFCD00038544

  •  PubChem Substance ID 24897758

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   OFYAYGJCPXRNBL-GFCCVEGCSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?