EMAIL THIS PAGE TO A FRIEND
N6388 Sigma

Neuromedin C

≥97% (HPLC)

Synonym: GRP (18-27)

  • CAS Number 81608-30-2

  • Empirical Formula (Hill Notation) C50H72N16O12S

  • Molecular Weight 1121.27

Purchase

Properties

assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C

Description

Amino Acid Sequence

Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2

Biochem/physiol Actions

Stimulates rat uterine smooth muscle

Other Notes

Bombesin-like peptide from porcine spinal cord

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. N6388.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?