• USA Home
  • N6395 - Immobilon®-NC, nitrocellulose Membranes

EMAIL THIS PAGE TO A FRIEND
N6395 Sigma

Immobilon®-NC, nitrocellulose Membranes

size 20 cm × 20 cm

Purchase

Properties

description   NC HAHY
sterility   sterile; autoclavable to 121 °C (use below 75 °C).
size   20 cm × 20 cm
pore size   0.45 μm

Description

General description

Manufactured for superior binding of biomolecules, outstanding mechanical strength, and low background.

Legal Information

HAHY and HATF are the Millipore Corp. designations for these products.

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

N5531 Biobond-Plus Nylon transfer membrane, sheet size 20 cm × 20 cm

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?