• USA Home
  • N6500 - β-Naphthoxyacetic acid

EMAIL THIS PAGE TO A FRIEND
N6500 Sigma

β-Naphthoxyacetic acid

practical grade, crystalline

Purchase

Properties

grade   practical grade
form   crystalline
color   gray

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AI9659000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

N3405 2-Naphthoxyacetic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?