• USA Home
  • N6520 - Immobilon®-NC HAHY membranes
EMAIL THIS PAGE TO A FRIEND
N6520 Sigma
Immobilon®-NC HAHY membranes
L × W 12 in. × 12 in.
Purchase
Description
General description
Nitrocellulose with a small amount of cellulose acetate for improved handling. Water-extractables <5%, including a Triton wetting agent. For Southern blots, protein electroblotting, capillary blotting of DNA, RNA, protein.
Legal Information
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Antibody Explorer
SNAP and Go!
Safety & Documentation
Safety Information
Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?