• USA Home
  • N6783 - N-(NBD-Aminocaproyl)sphingosine β-D-glucosyl

EMAIL THIS PAGE TO A FRIEND
N6783 Sigma

N-(NBD-Aminocaproyl)sphingosine β-D-glucosyl

>98%

Synonym: C6-NBD Sphingosine, beta-D-glucosyl, N-[6-[(7-Nitro-2-1,3-benzoxadiazol-4-yl)amino]caproyl]-D-glucosyl-β1-1′-sphingosine

  • Empirical Formula (Hill Notation) C36H59N5O11

  • Molecular Weight 737.88

  •  MDL number MFCD03453620

Purchase

Description

General description

Fluorescently labeled fatty acid sphingolipid membrane probe. NBD group excitation 460 nm, emission 534 nm.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?