• USA Home
  • N7139 - Naphthol AS-E acetate

EMAIL THIS PAGE TO A FRIEND
N7139 Sigma

Naphthol AS-E acetate

Synonym: 2-Naphthalenecarboxamide, 3-(acetyloxy)-N-(4-chlorophenyl)-, Azoic coupling component 10

  • CAS Number 84100-15-2

  • Empirical Formula (Hill Notation) C19H14ClNO3

  • Molecular Weight 339.77

  •  Colour Index Number 37510

  •  EC Number 282-097-1

  •  MDL number MFCD00042719

  •  PubChem Substance ID 24897812

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, N, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   INHDDLDQONYNHO-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?