• USA Home
  • N7268 - Nordoxepin hydrochloride

EMAIL THIS PAGE TO A FRIEND
N7268 Sigma

Nordoxepin hydrochloride

Synonym: 11(6H)-(3-[Methylamino]propylidene)dibenz[b,e]oxepine

  • CAS Number 2887-91-4

  • Empirical Formula (Hill Notation) C18H19NO · HCl

  • Molecular Weight 301.81

  •  MDL number MFCD00135797

  •  PubChem Substance ID 329818670

Purchase

Properties

InChI Key   GNPPEZGJRSOKRE-QFHYWFJHSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Tricyclic Antidepressants on Ascentis® Express 2.7 μm RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Tricyclic Antidepressants on Ascentis® Express 2.7 μm RP-Amide
Keywords: Anti-depressants, Chromatography, High performance liquid chromatography, Pharmaceutical, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?