• USA Home
  • N7270 - Immobilon®-NC HATF membranes

EMAIL THIS PAGE TO A FRIEND
N7270 Sigma

Immobilon®-NC HATF membranes

diam. 85 mm

Purchase

Properties

Related Categories Blotting Membranes and Paper, Electrophoresis Supplies, Labware, Manufacturer Browser, Millipore,
mfr. no.   Millipore, HATF08550
diam.   85 mm
pore size   0.45 μm

Description

General description

Nitrocellulose with a small amount of cellulose acetate for improved handling. Triton-free. For colony hybridization, plaque lifts, colony growth directly on membrane.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?