• USA Home
  • N7284 - Northern Bound Human Lung Tumor mRNA Blot
EMAIL THIS PAGE TO A FRIEND
N7284 Sigma
Northern Bound Human Lung Tumor mRNA Blot
Purchase
Description
General description
These blots contain high quality poly A+ RNA isolated from lung tumor and normal lung tissue. Lanes 1-3 contain mRNA isolated from lung tumor from three different donors. Lane 4 contains mRNA isolated from normal lung tissue. Lane 5 is an interblot control.
Legal Information
Manufactured by BioChain, Inc.
Northern Bound is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?