• USA Home
  • N7397 - Immobilon®-NC Pure Nitrocellulose Membranes
EMAIL THIS PAGE TO A FRIEND
N7397 Sigma
Immobilon®-NC Pure Nitrocellulose Membranes
Purchase
Properties
size   15 cm × 15 cm
pore size   0.22 μm
Description
General description
100% nitrocellulose with surfactant, for general blotting procedures; more flexible than standard NC membranes without reinforcing web
Legal Information
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
P2563 Immobilon®-P Polyvinylidene difluoride membranes, size 15 cm × 15 cm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?