• USA Home
  • N7641 - Hamilton® Microliter 7000 series syringe
EMAIL THIS PAGE TO A FRIEND
N7641 Sigma
Hamilton® Microliter 7000 series syringe
Needle with knurled hub for use with Model 7105KH, needle size 24 gauge
Purchase
Properties
Related Categories Hamilton syringes, Labware, Microliter, Syringes and Needles
needle size   24 gauge
Description
General description
Designed for rapid injection of minute volumes. The total syringe capacity is in the needle. The PTFE needle ferrule can be tightened to ensure a leakproof seal at injection pressures up to 6,000 psi.
Point style 3, blunt tip, needle length is 7 cm.
It is recommended that a chaney adapter be used with this syringe.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microliter is a trademark of Hamilton Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?