• USA Home
  • N7784 - β-Nicotinamide adenine dinucleotide, reduced disodium salt

EMAIL THIS PAGE TO A FRIEND
N7784 Sigma

β-Nicotinamide adenine dinucleotide, reduced disodium salt

Synonym: β-DPNH, β-NADH, Coenzyme I reduced disodium salt, Diphosphopyridine nucleotide reduced disodium salt

  • CAS Number 606-68-8

  • Empirical Formula (Hill Notation) C21H27N7Na2O14P2

  • Molecular Weight 709.40

  •  Beilstein Registry Number 5230241

  •  MDL number MFCD00036200

Purchase

Properties

InChI Key   QRGNQKGQENGQSE-QUWMEQBESA-L

Description

Biochem/physiol Actions

Electron donor

Other Notes

Packaged based on NADH content as determined by UV-Vis.

This is the common form of NADH; do not confuse with α-NADH.

Reconstitution

Solutions should be prepared fresh and used promptly.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?