MDL number MFCD00081525
PubChem Substance ID 24897844
| Related Categories | Blotting Membranes and Paper, Core Bioreagents, Electrophoresis Supplies, Equipment, Labware, |
| size | 15 cm × 15 cm |
| pore size | 0.2 μm |
Nitrocellulose membranes are the most popular membranes for immunoblotting (Western blotting) of proteins. The basis of protein binding to nitrocellulose is believed to be due to hydrophobic interactions. The 0.45 μm pore size is the most commonly used for proteins greater than 10,000 MW, but for smaller proteins 0.2 μm is recommended to more efficiently trap the proteins.1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?