• USA Home
  • N7908 - Monoclonal Anti-Nicotinic Acetylcholine Receptor (α5 Subunit) antibody produced in rat

EMAIL THIS PAGE TO A FRIEND
N7908 Sigma

Monoclonal Anti-Nicotinic Acetylcholine Receptor (α5 Subunit) antibody produced in rat

~5 mg/mL, clone mAb 268, purified immunoglobulin, buffered aqueous solution

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Neurotransmission, Antibodies to Ion Channels,
species reactivity   human, chicken
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  western blot: suitable
clone   mAb 268, monoclonal
concentration   ~5 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1/IgG2a
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CHRNA5(1138)
biological source   rat
conjugate   unconjugated

Description

Immunogen

nicotinic acetylcholine receptors from purified chick brain.

Physical form

Solution in 10 mM phosphate buffered saline, containing 10 mM sodium azide.

Specificity

Recognizes an epitope with the sequence 91-100 of the α5 subunit of nicotinic acetylcholine receptor. Binds to denatured α5 subunit of the nicotinic acetylcholine receptor. Does not bind to native α5 subunits.

Legal Information

Sold with the permission of the Salk Institute.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?