• USA Home
  • N8017 - Nitrocellulose membranes for blotting

EMAIL THIS PAGE TO A FRIEND
N8017 Sigma

Nitrocellulose membranes for blotting

pore size 0.2 μm, size 20 cm × 20 cm

Purchase

Description

Application

Nitrocellulose membranes are the most popular membranes for immunoblotting (Western blotting) of proteins. The basis of protein binding to nitrocellulose is believed to be due to hydrophobic interactions. The 0.45 μm pore size is the most commonly used for proteins greater than 10,000 MW, but for smaller proteins 0.2 μm is recommended to more efficiently trap the proteins.

Price and Availability


Detect Your Unseen Protein

Amicon Pro Purification System

SNAP and Go!
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?