• USA Home
  • N8022 - Immobilon®-S Nylon membranes
EMAIL THIS PAGE TO A FRIEND
N8022 Sigma
Immobilon®-S Nylon membranes
sheet diam. 82 mm
Purchase
Properties
Related Categories Blotting Membranes and Paper, Electrophoresis Supplies, Labware, Manufacturer Browser, Millipore,
mfr. no.   Millipore, INYC08250
sheet diam.   82 mm
pore size   0.45 μm
Description
General description
Uncharged transfer membrane for isotopic or chemiluminescent detection of nucleic acids.
Easily cross-linked and reprobed.
Legal Information
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?