EMAIL THIS PAGE TO A FRIEND
N8125 Sigma

2-Naphthyl butyrate

Synonym: Butanoic acid 2-naphthyl ester

  • CAS Number 5856-33-7

  • Empirical Formula (Hill Notation) C14H14O2

  • Molecular Weight 214.26

  •  MDL number MFCD00046464

  •  PubChem Substance ID 24897852

Purchase

Properties

Related Categories Biochemicals and Reagents, Chromogen, Enzyme Substrate, Fatty Acyls, Lipids More...
InChI Key   KCWQXDQUKLJUEU-UHFFFAOYSA-N
density   1.12 g/mL at 25 °C(lit.)
storage temp.   −20°C

Description

Packaging

Sealed ampule.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?