• USA Home
  • N8154 - [Ala31, α-Aminoisobutyryl32]-Neuropeptide Y porcine

EMAIL THIS PAGE TO A FRIEND
N8154 Sigma

[Ala31, α-Aminoisobutyryl32]-Neuropeptide Y porcine

≥98% (HPLC), solid

Synonym: [Ala31, (α-Methyl-Ala)32]-Neuropeptide Y porcine, [Ala31, Aib32]-NPY Porcine

  • Empirical Formula (Hill Notation) C187H281N55O56

  • Molecular Weight 4195.57

  •  MDL number MFCD03096323

Purchase

Description

Amino Acid Sequence

Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ala-alpha-aminoisobutyryl (α-Methyl-Ala-Arg-Gln-Arg-Tyr-NH2)

Biochem/physiol Actions

Agonist for the neuropeptide Y5 receptor exhibiting high affinity and selectivity.

Price and Availability


Amplified Detection

Making Headway

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?