EMAIL THIS PAGE TO A FRIEND
N8250 Sigma

1-Naphthyl decanoate

Synonym: Capric acid 1-naphthyl ester

Purchase

Properties

Related Categories Biochemicals and Reagents, Chromogen, Enzyme Substrate, Fatty Acyls, Lipids More...
InChI Key   BHZDUUIJMMLTMB-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

N0252 4-Nitrophenyl decanoate, lipase substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?