• USA Home
  • N8284 - N-(NBD-Aminolauroyl)phytosphingosine

EMAIL THIS PAGE TO A FRIEND
N8284 Sigma

N-(NBD-Aminolauroyl)phytosphingosine

>98%

Synonym: C12-NBD Phytosphingosine, N-[12-[(7-Nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-phytosphingosine

  • Empirical Formula (Hill Notation) C36H63N5O7

  • Molecular Weight 677.91

  •  MDL number MFCD02259243

  •  PubChem Substance ID 329818684

Purchase

Description

General description

Fluorescently labeled fatty acid sphingolipid membrane probe. NBD group excitation 460 nm, emission 534 nm.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?