• USA Home
  • N8647 - Immobilon®-S Nylon membranes
EMAIL THIS PAGE TO A FRIEND
N8647 Sigma
Immobilon®-S Nylon membranes
roll W × L 30 cm × 3.5 m
Purchase
Properties
Related Categories Blotting Membranes and Paper, Electrophoresis Supplies, Labware, Manufacturer Browser, Millipore,
mfr. no.   Millipore, INYC00010
roll W × L   30 cm × 3.5 m
pore size   0.45 μm
Description
General description
Uncharged transfer membrane for isotopic or chemiluminescent detection of nucleic acids.
Easily cross-linked and reprobed.
Legal Information
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?