• USA Home
  • N8777 - β-Nicotinamide adenine dinucleotide phosphate disodium salt

EMAIL THIS PAGE TO A FRIEND
N8777 Sigma

β-Nicotinamide adenine dinucleotide phosphate disodium salt

Synonym: β-NADH phosphate disodium salt, β-NADP-Na2, Triphosphopyridine nucleotide disodium salt

  • CAS Number 24292-60-2

  • Empirical Formula (Hill Notation) C21H26N7Na2O17P3

  • Molecular Weight 787.37

  •  MDL number MFCD00065390

  •  PubChem Substance ID 329818689

Purchase

Properties

InChI Key   WSDDJLMGYRLUKR-FHQWUTKFSA-L

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?