• USA Home
  • N8783 - Monoclonal Anti-Nicotinic Acetylcholine Receptor (δ Subunit) antibody produced in rat

EMAIL THIS PAGE TO A FRIEND
N8783 Sigma

Monoclonal Anti-Nicotinic Acetylcholine Receptor (δ Subunit) antibody produced in rat

~5 mg/mL, clone mAb 166, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Neurotransmission, Antibodies to Ion Channels,
biological source   rat
antibody form   purified immunoglobulin
clone   mAb 166, monoclonal
form   buffered aqueous solution
species reactivity   amphibian, Torpedo
concentration   ~5 mg/mL
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  western blot: suitable
isotype   IgG2a
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Immunogen

nicotinic acetylcholine receptors from the electric eel Torpedo.

Legal Information

Sold with the permission of the Salk Institute.

Specificity

Recognizes δ subunits of Torpedo acetylcholine receptors at an epitope within amino acids 300-410. May be used with native and denatured Torpedo tissues and amphibian muscle, but is not useful for mammalian δ tissues.

Physical form

Solution in 10 mM phosphate buffered saline, pH 7.4, containing 10 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?