• USA Home
  • N8882 - NAD+ Kinase from chicken liver

EMAIL THIS PAGE TO A FRIEND
N8882 Sigma

NAD+ Kinase from chicken liver

Type IV, lyophilized powder, 10-20 units/mg solid

Synonym: ATP: NAD+ 2′-phosphotransferase

Purchase

Properties

Related Categories 2.7.x.x Phosphorus containing groups, 2.x.x.x Transferases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
type   Type IV
form   lyophilized powder
composition   Protein, ≥80% biuret
storage temp.   −20°C
Gene Information   chicken ... RCJMB04_9i6(419403)

Description

Biochem/physiol Actions

Reaction: β-NAD + ATP → β-NADP + ADP.

Unit Definition

One unit will phosphorylate 1.0 nanomole of β-NAD to β-NADP per min at pH 7.5 at 37 °C, in the presence of ATP.

Physical form

Lyophilized powder containing citrate buffer

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?