• USA Home
  • N9270 - Filter membranes, nitrocellulose

EMAIL THIS PAGE TO A FRIEND
N9270 Sigma

Filter membranes, nitrocellulose

MF membrane, pore size 0.45 μm, diam. 142 mm

Purchase

Properties

Related Categories Filter membranes, nitrocellulose, Filtration Supplies, Labware, Manufacturer Browser, Membrane Filters,
material   MF membrane
mfr. no.   Millipore HAWP14250
  Millipore, HAWP14250
diam.   142 mm
pore size   0.45 μm

Description

General description

Biologically inert nitrocellulose membranes containing a small amount of cellulose acetate for improved handling. Autoclavable to 121°C; use below 75°C. Chemically compatible with dilute acids and bases, hydrocarbons, and non-polar liquids. MF-Millipore membranes have water extractables <5%, including a Triton wetting agent TF-Millipore membranes are Triton-free and have even lower water extractables than MF membranes. See Immobilon-NC in the Electrophoresis Equipment section for nitrocellulose blotting membranes.

Price and Availability


Sterifil Aseptic System and Holder
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?